4-(2,2-Dichloroacetyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

83143-1G 83143-500MG 83143-5G 83143-25G
77473-456EA 415.09 USD
77473-456 77486-072 77496-686 77505-696
4-(2,2-Dichloroacetyl)benzonitrile ≥95%
4-(2,2-Dichloroacetyl)benzonitrile

Precaución: For research use only

Formula: C₉H₅Cl₂NO
Storage Temperature: Ambient
CAS Number: 35747-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR