2,2-Dichloro-1-(4-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

83028-1G 83028-500MG 83028-5G 83028-25G
77473-452EA 347.11 USD
77473-452 77486-068 77496-682 77505-692
2,2-Dichloro-1-(4-fluorophenyl)ethanone ≥95%
2,2-Dichloro-1-(4-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₅Cl₂FO
MW: 207.029 g/mol
Storage Temperature: Ambient
CAS Number: 5157-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR