1-(2-Chloropyridin-3-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

82938-1G 82938-500MG 82938-5G
77473-448EA 557.23 USD
77473-448 77486-064 77496-676
1-(2-Chloropyridin-3-yl)-2,2-difluoroethanone ≥97%
1-(2-Chloropyridin-3-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₄ClF₂NO
MW: 191.563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13172774
CAS Number: 335449-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR