2-(3-Bromo-5-fluoro-2-methoxyphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

82835-1G 82835-500MG 82835-5G 82835-25G
77473-444EA 374.92 USD
77473-444 77486-060 77496-670 77505-682
2-(3-Bromo-5-fluoro-2-methoxyphenyl)propan-2-ol ≥97%
2-(3-Bromo-5-fluoro-2-methoxyphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769366
CAS Number: 1785005-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR