Ethyl 2-(4-(difluoromethoxy)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82771-1G 82771-500MG 82771-5G
77473-440EA 576.8 USD
77473-440 77486-056 77496-666
Ethyl 2-(4-(difluoromethoxy)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-(difluoromethoxy)phenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₁H₁₀F₂O₄
MW: 244.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22688404
CAS Number: 1417460-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR