Ethyl 2-(4-bromo-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82628-1G 82628-500MG 82628-5G
77473-432EA 442.9 USD
77473-432 77486-048 77496-658
Ethyl 2-(4-bromo-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-bromo-3-methylphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₁H₁₁BrO₃
MW: 271.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28106989
CAS Number: 1368032-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR