Ethyl 2-oxo-2-(2-(trifluoromethoxy)phenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

82560-1G 82560-500MG 82560-5G 82560-25G
77473-428EA 266.77 USD
77473-428 77486-044 77496-654 77505-676
Ethyl 2-oxo-2-(2-(trifluoromethoxy)phenyl)acetate ≥95%
Ethyl 2-oxo-2-(2-(trifluoromethoxy)phenyl)acetate

Precaución: For research use only

Formula: C₁₁H₉F₃O₄
MW: 262.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370289
CAS Number: 334778-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR