Ethyl 2-(3-(tert-butyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82543-1G 82543-500MG 82543-5G 82543-25G
77473-424EA 264.71 USD
77473-424 77486-040 77496-650 77505-672
Ethyl 2-(3-(tert-butyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-(tert-butyl)phenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₄H₁₈O₃
MW: 234.291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113520
CAS Number: 1256481-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR