Ethyl 2-(4-(methoxymethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82504-1G 82504-500MG 82504-5G 82504-25G
77473-420EA 337.84 USD
77473-420 77486-036 77496-646 77505-670
Ethyl 2-(4-(methoxymethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-(methoxymethyl)phenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₂H₁₄O₄
MW: 222.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11555689
CAS Number: 1256466-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR