Ethyl-2,5-dimethoxybenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82479-1G 82479-500MG 82479-5G
77473-416EA 436.72 USD
77473-416 77486-032 77496-642
Ethyl-2,5-dimethoxybenzoylformate ≥95%
Ethyl-2,5-dimethoxybenzoylformate

Precaución: For research use only

Formula: C₁₂H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD09801405
CAS Number: 911047-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR