Ethyl-2,3-dimethylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

82477-1G 82477-500MG 82477-5G 82477-25G
77473-412EA 226.6 USD
77473-412 77486-028 77496-638 77505-664
Ethyl-2,3-dimethylbenzoylformate ≥95%
Ethyl-2,3-dimethylbenzoylformate

Precaución: For research use only

Formula: C₁₂H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD09801407
CAS Number: 951888-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR