Ethyl-3-fluoro-2-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-fluoro-2-methylphenyl)-2-oxoacetate

82466-1G 82466-500MG 82466-5G 82466-25G
77473-408EA 267.8 USD
77473-408 77486-024 77496-634 77505-660
Ethyl-3-fluoro-2-methylbenzoylformate ≥95%
Ethyl-3-fluoro-2-methylbenzoylformate

Precaución: For research use only

Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801424
CAS Number: 951888-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR