Ethyl 2-(2,4-dimethylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82456-1G 82456-500MG 82456-5G 82456-25G
77473-404EA 217.33 USD
77473-404 77486-020 77496-630 77505-656
Ethyl 2-(2,4-dimethylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,4-dimethylphenyl)-2-oxoacetate

Precaución: For research use only

Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801408
CAS Number: 80120-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR