Ethyl-4-chloro-2-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(4-chloro-2-fluorophenyl)-2-oxoacetate

82433-1G 82433-500MG 82433-5G 82433-25G
77473-400EA 279.13 USD
77473-400 77486-016 77496-626 77505-652
Ethyl-4-chloro-2-fluorobenzoylformate ≥95%
Ethyl-4-chloro-2-fluorobenzoylformate

Precaución: For research use only

Formula: C₁₀H₈ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD09801394
CAS Number: 228998-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR