Ethyl-2,4-difluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2,4-difluorophenyl)-2-oxoacetate

82411-1G 82411-500MG 82411-5G 82411-25G
77473-396EA 296.64 USD
77473-396 77486-012 77496-620 77505-646
Ethyl-2,4-difluorobenzoylformate ≥95%
Ethyl-2,4-difluorobenzoylformate

Precaución: For research use only

Formula: C₁₀H₈F₂O₃
MW: 214.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205165
CAS Number: 157372-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR