Ethyl-2-methylbenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-oxo-2-(o-tolyl)acetate

82404-1G 82404-500MG 82404-5G 82404-25G
77473-392EA 209.09 USD
77473-392 77486-008 77496-616 77505-642
Ethyl-2-methylbenzoylformate ≥95%
Ethyl-2-methylbenzoylformate

Precaución: For research use only

Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01311167
CAS Number: 66644-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR