1-(4-Bromo-2-methylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

82388-1G 82388-500MG 82388-5G
77473-388EA 486.16 USD
77473-388 77486-004 77496-612
1-(4-Bromo-2-methylphenyl)cyclohexanol ≥95%
1-(4-Bromo-2-methylphenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₃H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD30477443
CAS Number: 2060044-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR