1-(4-Bromo-2,5-dimethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

82382-1G 82382-500MG 82382-5G
77473-384EA 546.93 USD
77473-384 77486-000 77496-608
1-(4-Bromo-2,5-dimethylphenyl)cyclobutanol ≥95%
1-(4-Bromo-2,5-dimethylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₂H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD34769308
CAS Number: 2901083-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR