(2-Bromo-3-(1,3-dioxolan-2-yl)-6-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

82242-1G 82242-500MG 82242-5G 82242-25G
77473-376EA 415.09 USD
77473-376 77485-992 77496-600 77505-630
(2-Bromo-3-(1,3-dioxolan-2-yl)-6-fluorophenyl)trimethylsilane ≥97%
(2-Bromo-3-(1,3-dioxolan-2-yl)-6-fluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₁₆BrFO₂Si
MW: 319.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34816840
CAS Number: 2921836-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR