2,6-Dichloro-4-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

82087-1G 82087-500MG 82087-5G 82087-25G
77473-372EA 204.97 USD
77473-372 77485-988 77496-596 77505-626
2,6-Dichloro-4-hydroxybenzoic acid ≥95%
2,6-Dichloro-4-hydroxybenzoic acid

Precaución: For research use only

Formula: C₇H₄Cl₂O₃
MW: 207.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24479951
CAS Number: 4641-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR