(3-Chloro-2,6-difluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

81816-1G 81816-500MG 81816-5G 81816-25G
77473-368EA 415.09 USD
77473-368 77485-984 77496-592 77505-624
(3-Chloro-2,6-difluorophenyl)(ethyl)sulfane ≥95%
(3-Chloro-2,6-difluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₇ClF₂S
Storage Temperature: Ambient
MDL Number: MFCD34816770
CAS Number: 2921716-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR