5-Bromo-2-(ethylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

81799-1G 81799-500MG 81799-5G 81799-25G
77473-360EA 122.57 USD
77473-360 77485-976 77496-584 77505-616
5-Bromo-2-(ethylthio)pyridine ≥95%
5-Bromo-2-(ethylthio)pyridine

Precaución: For research use only

Formula: C₇H₈BrNS
MW: 218.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14650168
CAS Number: 127800-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR