5-Bromo-2-fluoro-3-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81441-1G 81441-500MG 81441-5G
77473-352EA 508.82 USD
77473-352 77485-968 77496-576
5-Bromo-2-fluoro-3-propoxybenzaldehyde ≥97%
5-Bromo-2-fluoro-3-propoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768802
CAS Number: 2901109-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR