1-(4-Methoxyphenyl)ethane-1,2-diol ≥97%

Supplier: AOBChem USA

Warning

81297-1G 81297-500MG 81297-5G 81297-25G
77473-348EA 355.35 USD
77473-348 77485-964 77496-568 77505-604
1-(4-Methoxyphenyl)ethane-1,2-diol ≥97%
1-(4-Methoxyphenyl)ethane-1,2-diol

Precaución: For research use only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521288
CAS Number: 13603-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR