(3-Chloro-6-fluoro-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

80963-1G 80963-500MG 80963-5G 80963-25G
77473-340EA 415.09 USD
77473-340 77485-956 77496-560 77505-602
(3-Chloro-6-fluoro-2-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(3-Chloro-6-fluoro-2-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀ClFO₂S
MW: 236.691 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768541
CAS Number: 2901102-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR