2-(Benzyloxy)-1-bromo-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

80953-1G 80953-500MG 80953-5G 80953-25G
77473-336EA 282.22 USD
77473-336 77485-952 77496-556 77505-598
2-(Benzyloxy)-1-bromo-3-methoxybenzene ≥95%
2-(Benzyloxy)-1-bromo-3-methoxybenzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33402460
CAS Number: 1996994-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR