1-(3-Bromo-2,4-dichloro-6-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

80742-1G 80742-500MG 80742-5G
77473-332EA 546.93 USD
77473-332 77485-948 77496-548
1-(3-Bromo-2,4-dichloro-6-fluorophenyl)ethanol ≥95%
1-(3-Bromo-2,4-dichloro-6-fluorophenyl)ethanol

Precaución: For research use only

Formula: C₈H₆BrCl₂FO
MW: 287.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768466
CAS Number: 2901099-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR