(2-Fluoro-5-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80613-1G 80613-500MG 80613-5G
77473-328EA 546.93 USD
77473-328 77485-944 77496-544
(2-Fluoro-5-methoxyphenyl)trimethylsilane ≥95%
(2-Fluoro-5-methoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₅FOSi
MW: 198.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34687559
CAS Number: 125889-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR