(2-Chloro-3-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

80567-1G 80567-500MG 80567-5G 80567-25G
77473-324EA 415.09 USD
77473-324 77485-940 77496-540 77505-588
(2-Chloro-3-(methoxymethoxy)phenyl)(methyl)sulfane ≥97%
(2-Chloro-3-(methoxymethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁ClO₂S
MW: 218.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768399
CAS Number: 2901105-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR