3-Bromo-2-ethoxy-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80487-1G 80487-500MG 80487-5G
77473-320EA 546.93 USD
77473-320 77485-936 77496-536
3-Bromo-2-ethoxy-5-formylbenzoic acid ≥97%
3-Bromo-2-ethoxy-5-formylbenzoic acid

Precaución: For research use only

Formula: C₁₀H₉BrO₄
MW: 273.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768349
CAS Number: 2248276-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR