(6-Chloro-2-fluoro-3-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80460-1G 80460-500MG 80460-5G 80460-25G
77473-316EA 415.09 USD
77473-316 77485-932 77496-532 77505-582
(6-Chloro-2-fluoro-3-methoxyphenyl)trimethylsilane ≥95%
(6-Chloro-2-fluoro-3-methoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₄ClFOSi
Storage Temperature: Ambient
MDL Number: MFCD31807988
CAS Number: 2918884-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR