(2-Chloro-6-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

80452-1G 80452-500MG 80452-5G 80452-25G
77473-312EA 111.24 USD
77473-312 77485-928 77496-528 77505-580
(2-Chloro-6-fluorophenyl)trimethylsilane ≥97%
(2-Chloro-6-fluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₂ClFSi
Storage Temperature: Ambient
MDL Number: MFCD27976481
CAS Number: 133117-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR