1-(2,6-Dibromophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

80444-1G 80444-500MG 80444-5G 80444-25G
77473-308EA 255.44 USD
77473-308 77485-924 77496-524 77505-578
1-(2,6-Dibromophenyl)ethanol ≥97%
1-(2,6-Dibromophenyl)ethanol

Precaución: For research use only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768320
CAS Number: 1675968-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR