5-Bromo-7-methoxy-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

80289-1G 80289-500MG 80289-5G 80289-25G
77473-300EA 91.67 USD
77473-300 77485-916 77496-516 77505-574
5-Bromo-7-methoxy-1H-indazole ≥95%
5-Bromo-7-methoxy-1H-indazole

Precaución: For research use only

Formula: C₈H₇BrN₂O
MW: 227.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11878570
CAS Number: 1374652-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR