5-Bromo-2-iodobenzo[b]thiophene ≥95%

Supplier: AOBChem USA

Warning

80024-1G 80024-500MG 80024-5G 80024-25G
77473-296EA 72.1 USD
77473-296 77485-912 77496-512 77505-572
5-Bromo-2-iodobenzo[b]thiophene ≥95%
5-Bromo-2-iodobenzo[b]thiophene

Precaución: For research use only

Formula: C₈H₄BrIS
Storage Temperature: Ambient
MDL Number: MFCD08706227
CAS Number: 306762-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR