5-Bromo-benzo[b]thiophene-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

80021-1G 80021-500MG 80021-5G 80021-25G
77473-292EA 110.21 USD
77473-292 77485-908 77496-508 77505-568
5-Bromo-benzo[b]thiophene-2-carbaldehyde ≥95%
5-Bromo-benzo[b]thiophene-2-carbaldehyde

Precaución: For research use only

Formula: C₉H₅BrOS
Storage Temperature: Ambient
MDL Number: MFCD11227154
CAS Number: 7312-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR