2-Bromo-4-(bromomethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

79973-1G 79973-5G 79973-25G
77473-288EA 70.04 USD
77473-288 77496-502 77505-562
2-Bromo-4-(bromomethyl)pyridine ≥95%
2-Bromo-4-(bromomethyl)pyridine

Precaución: For research use only

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847700
CAS Number: 83004-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR