5-Isopropoxy-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

79720-1G 79720-500MG 79720-5G 79720-25G
77473-284EA 163.77 USD
77473-284 77485-902 77496-498 77505-558
5-Isopropoxy-2-methoxybenzaldehyde ≥97%
5-Isopropoxy-2-methoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29121440
CAS Number: 1289047-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR