1-(2-Chloro-3,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

79585-1G 79585-500MG 79585-5G
77473-280EA 597.4 USD
77473-280 77485-898 77496-494
1-(2-Chloro-3,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Chloro-3,6-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₈H₂ClF₅O
MW: 244.546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922614
CAS Number: 1208077-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR