2,2-Difluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

79561-1G 79561-500MG 79561-5G 79561-25G
77473-272EA 415.09 USD
77473-272 77485-890 77496-486 77505-548
2,2-Difluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone ≥95%
2,2-Difluoro-1-(2,3,5-trichloropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₂Cl₃F₂NO
MW: 260.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34816329
CAS Number: 2918963-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR