1-(5-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

79542-1G 79542-500MG 79542-5G
77473-268EA 557.23 USD
77473-268 77485-886 77496-482
1-(5-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone ≥97%
1-(5-Bromo-2-chloropyridin-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₃BrClF₂NO
MW: 270.459 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34816327
CAS Number: 2228469-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR