tert-Butyl 3-chloroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

79527-1G 79527-500MG 79527-5G
77473-264EA 504.7 USD
77473-264 77485-882 77496-478
tert-Butyl 3-chloroisonicotinate ≥97%
tert-Butyl 3-chloroisonicotinate

Precaución: For research use only

Formula: C₁₀H₁₂ClNO₂
MW: 213.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18910175
CAS Number: 1466869-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR