(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one ≥95%

Supplier: AOBChem USA

Warning

79506-1G 79506-500MG 79506-5G 79506-25G
77473-260EA 151.41 USD
77473-260 77485-878 77496-474 77505-542
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one ≥95%
(2E)-1-(3-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one

Precaución: For research use only

Formula: C₉H₁₀BrNOS
Storage Temperature: Ambient
MDL Number: MFCD03618050
CAS Number: 1422045-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR