2,5-Diiodo-3-methylthiophene ≥97%
Supplier: AOBChem USA
|
Warning
|
79450-1G
79450-500MG
79450-5G
79450-25G
77473-252EA
210.12
USD
77473-252
77485-870
77496-466
77505-536
2,5-Diiodo-3-methylthiophene ≥97%
2,5-Diiodo-3-methylthiophene
Precaución: For research use only
Formula:
C₅H₄I₂S MW: 349.959 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11870143 CAS Number: 16488-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...