5-Iodo-3-methyl-2-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

79445-1G 79445-500MG 79445-5G 79445-25G
77473-248EA 415.09 USD
77473-248 77485-866 77496-462 77505-532
5-Iodo-3-methyl-2-(methylthio)thiophene ≥97%
5-Iodo-3-methyl-2-(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₇IS₂
MW: 270.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767979
CAS Number: 2901105-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR