4-Methyl-5-(methylthio)thiophene-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

79443-1G 79443-500MG 79443-5G 79443-25G
77473-244EA 415.09 USD
77473-244 77485-862 77496-458 77505-528
4-Methyl-5-(methylthio)thiophene-2-carboxylic acid ≥97%
4-Methyl-5-(methylthio)thiophene-2-carboxylic acid

Precaución: For research use only

Formula: C₇H₈O₂S₂
MW: 188.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767977
CAS Number: 2901085-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR