3-Methyl-2-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

79441-1G 79441-500MG 79441-5G 79441-25G
77473-240EA 339.9 USD
77473-240 77485-858 77496-454 77505-524
3-Methyl-2-(methylthio)thiophene ≥95%
3-Methyl-2-(methylthio)thiophene

Precaución: For research use only

Formula: C₆H₈S₂
MW: 144.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617754
CAS Number: 84034-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR