5-Iodo-4-methylthiophene-2-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

79439-1G 79439-500MG 79439-5G 79439-25G
77473-236EA 415.09 USD
77473-236 77485-854 77496-450 77505-520
5-Iodo-4-methylthiophene-2-carbaldehyde ≥97%
5-Iodo-4-methylthiophene-2-carbaldehyde

Precaución: For research use only

Formula: C₆H₅IOS
Storage Temperature: Ambient
MDL Number: MFCD34767975
CAS Number: 2901095-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR