5-Formyl-3-methylthiophene-2-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
79437-1G
79437-500MG
79437-5G
79437-25G
77473-232EA
307.97
USD
77473-232
77485-850
77496-446
77505-516
5-Formyl-3-methylthiophene-2-carboxylic acid ≥97%
5-Formyl-3-methylthiophene-2-carboxylic acid
Precaución: For research use only
Formula:
C₇H₆O₃S Storage Temperature: Ambient |
MDL Number:
MFCD22067241 CAS Number: 1368183-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...