2-(3-Bromo-2,5-difluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

79415-1G 79415-500MG 79415-5G 79415-25G
77473-228EA 415.09 USD
77473-228 77485-846 77496-442 77505-512
2-(3-Bromo-2,5-difluorophenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-2,5-difluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767969
CAS Number: 2901109-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR